Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP28 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550237

Catalog No. PA550237


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total MMP28 protein.

Epilysin, also known as matrix metalloproteinase-28 (MMP-28), is a secreted protein that belongs to the peptidase M10A family. Matrix metalloproteinase proteins are responsible for the breakdown of the extracellular matrix which is important for normal physiological processes such as tissue remodeling, reproduction and embryonic development. Epilysin is produced by proliferating keratinocytes and is responsible for mediating the degradation of casein. Its expression is upregulated in response to injury of the skin suggesting that Epilysin may be involved in tissue repair and homeostasis. In addition, Epilysin may function predominantly in the nervous system. Incubation of Epilysin with rat embryonic brain tissue results in the degradation of myelin proteins. In Xenopus, Epilysin localizes to nerves and is upregulated during neurulation. This suggests that Epilysin plays an evolutionarily conserved role in neural development and may be involved in modulating the axonal-glial extracellular environment.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP28
Polyclonal
Unconjugated
MMP28
D130023P12; Epilysin; matrix metallo protease; matrix metallopeptidase 28; matrix metallopeptidase 28 (epilysin); matrix metalloprotease MMP25; matrix metalloproteinase 28 (epilysin); matrix metalloproteinase-28; MM28; MMP; MMP25; MMP-25; Mmp28; MMP-28; MMPs; PRO4339; UNQ1893; UNQ1893/PRO4339
Rabbit
Antigen affinity chromatography
RUO
118453, 79148
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9H239
MMP28
Fusion protein corresponding to residues near the C terminal of human matrix metallopeptidase 28.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.