Learn More
MMP3 Mouse anti-Human, Clone: OTI2D7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806933

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifications
MMP3 | |
Monoclonal | |
Unconjugated | |
MMP3 | |
CHDS6, MMP-3, SL-1, STMY, STMY1, STR1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4314 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08254 | |
MMP3 | |
Human recombit protein fragment corresponding to amino acids 246-477 of human MMP3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.