Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP3 Rabbit anti-Human, Unconjugated, Clone: MMP3/1994R, Abnova™

Catalog No. 89337945 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-945 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-945 Supplier Abnova Corporation Supplier No. RAB00520
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

MMP3 Monoclonal antibody specifically detects MMP3 in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen MMP3
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone MMP3/1994R
Conjugate Unconjugated
Description matrix metallopeptidase 3 (stromelysin 1, progelatinase)
Gene MMP3
Gene Alias CHDS6, MGC126102, MGC126103, MGC126104, MMP-3, SL-1, STMY, STMY1, STR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 317-327 of human MMP3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4314
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.