Learn More
Invitrogen™ MMP7 Polyclonal Antibody

Description
Positive Control: A549 cell lysates, HT-29 cell lysate, A431 cell lysate, A549, human uterus tissue, HT-29. Subcellular Location: Extracellular matrix, Secreted.
Specifications
Specifications
| Antigen | MMP7 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | MMP7 |
| Gene Accession No. | P09237 |
| Gene Alias | HGNC:7174; MAT; Matrilysin; matrilysin, uterine; matrin; matrix metallo protease; matrix metallopeptidase 7; matrix metallopeptidase 7 (matrilysin, uterine); matrix metalloproteinase 7; Matrix metalloproteinase 7 (matrilysin); matrix metalloproteinase 7 (matrilysin, uterine); matrix metalloproteinase-7; MMP; MMP 7; MMP7; Mmp-7; MMP-7 protein; MMPs; MPMM; MPSL1; PUMP 1; Pump 1 protease; PUMP1; PUMP-1; pump-1 protease; uterine matrilysin; uterine metalloproteinase |
| Gene Symbols | MMP7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.