Learn More
MMP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 178741AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-1847
Only null left
Add to Cart
Description
Description
Can degrade fibrillar type I, II, and III collagens.
Specifications
Specifications
MMP8
Polyclonal
Unconjugated
MMP8
CLG1, HNC, Matrix metalloproteinase 8, MMP 8, MMP8, Neutrophil collagenase, PMNL CL, PMNL collagenase
Rabbit
Antigen Affinity Chromatography
RUO
17394, 4317, 63849
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70138, O88766, P22894
Mmp8
MMP8 Fusion Protein Ag12367
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.