Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP9 Mouse anti-Human, Unconjugated, Clone: rMMP9/1769, Abnova™

Catalog No. 89337956 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-956 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-956 Supplier Abnova Corporation Supplier No. RAB00531
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MMP9 Monoclonal antibody specifically detects MMP9 in Human samples. It is validated for IHC-P.

Specifications

Antigen MMP9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMMP9/1769
Conjugate Unconjugated
Description matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase)
Gene MMP9
Gene Alias CLG4B, GELB, MMP-9
Host Species Mouse
Immunogen Recombinant protein corresponding to full-length human MMP9
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4318
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.