Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMRN1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548816

Catalog No. PA548816


Only null left
Add to Cart

Description

Description

MMRN1 Polyclonal Antibody for Western Blot

Multimerin-1, also known as MMRN1, EMILIN-4 or ECM (endothelial cell multimerin), is a 1,228 amino acid secreted protein that contains one C1q domain, one EMI domain and one EGF-like domain. Synthesized in megakaryocytes and endothelial cells and present in liver, lung and placenta, Multimerin-1 exists as a multimeric structure composed of varying disulfide-linked multimers and functions as a carrier protein for platelet factors (specifically platelet factor V), playing a role in the stabilization and storage of factor V in platelets. In addition, Multimerin-1 acts as a ligand for select Integrins and may participate in extracellular matrix adhesion. Defects in the gene encoding Multimerin-1 that lead to Multimerin-1 deficiency are associated with autosomal dominant bleeding disorders due to platelet factor malfunction. Multiple isoforms of Multimerin-1 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMRN1
Polyclonal
Unconjugated
MMRN1
155 kDa platelet multimerin; 4921530G03Rik; ECM; Elastin microfibril interface located protein 4; elastin microfibril interfacer 4; EMILIN4; EMILIN-4; Endothelial cell multimerin; glycoprotein Ia*; GPIa*; MMRN; mmrn1; multimerin 1; multimerin-1; p155; p-155; Platelet glycoprotein Ia*
Rabbit
Antigen affinity chromatography, Protein A
RUO
22915
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q13201
MMRN1
KLH conjugated synthetic peptide between 179-207 amino acids from the N-terminal region of human MMRN1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.