Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMS19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100071
Description
Antibody detects endogenous levels of total MMS19.
Key component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into apoproteins specifically involved in DNA metabolism and genomic integrity. In the CIA complex, MMS19 acts as an adapter between early-acting CIA components and a subset of cellular target iron-sulfur proteins such as ERCC2/XPD, FANCJ and RTEL1, thereby playing a key role in nucleotide excision repair (NER) and RNA polymerase II (POL II) transcription. As part of the mitotic spindle-associated MMXD complex, plays a role in chromosome segregation, probably by facilitating iron-sulfur cluster assembly into ERCC2/XPD. Indirectly acts as a transcriptional coactivator of estrogen receptor (ER), via its role in iron-sulfur insertion into some component of the TFIIH-machinery.
Specifications
MMS19 | |
Polyclonal | |
Unconjugated | |
MMS19 | |
2410001K24Rik; 2610042O15Rik; AI316855; C79368; C86341; hMMS19; homolog of yeast MMS19; incisor protein; MET18; MET18 homolog; Mms19; MMS19 (MET18 S. cerevisiae); MMS19 cytosolic iron-sulfur assembly component; MMS19 homolog, cytosolic iron-sulfur assembly component; MMS19 nucleotide excision repair homolog; MMS19 nucleotide excision repair homolog (S. cerevisiae); MMS19 nucleotide excision repair protein homolog; MMS19 nucleotide excision repair-like; Mms19l; MMS19-like (MET18 homolog, S. cerevisiae); MMS19-like protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
171124, 64210, 72199 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96T76, Q9D071 | |
MMS19 | |
A synthesized peptide derived from human MMS19(Accession Q96T76), corresponding to amino acid residues E342-N392. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction