Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMS19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584093 Supplier Invitrogen™ Supplier No. PA584093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVFMALTDPS TQLQLVGIRT LTVLGAQPDL LSYEDLELAV GHLYRLSFLK EDSQSCRVAA LEASGTLAAL YPVAFSSHLV PKLA Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 88%.

Key component of the cytosolic iron-sulfur protein assembly (CIA) complex, a multiprotein complex that mediates the incorporation of iron-sulfur cluster into apoproteins specifically involved in DNA metabolism and genomic integrity. In the CIA complex, MMS19 acts as an adapter between early-acting CIA components and a subset of cellular target iron-sulfur proteins such as ERCC2/XPD, FANCJ and RTEL1, thereby playing a key role in nucleotide excision repair (NER) and RNA polymerase II (POL II) transcription. As part of the mitotic spindle-associated MMXD complex, plays a role in chromosome segregation, probably by facilitating iron-sulfur cluster assembly into ERCC2/XPD. Indirectly acts as a transcriptional coactivator of estrogen receptor (ER), via its role in iron-sulfur insertion into some component of the TFIIH-machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMS19
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MMS19
Gene Accession No. Q96T76
Gene Alias 2410001K24Rik; 2610042O15Rik; AI316855; C79368; C86341; hMMS19; homolog of yeast MMS19; incisor protein; MET18; MET18 homolog; Mms19; MMS19 (MET18 S. cerevisiae); MMS19 cytosolic iron-sulfur assembly component; MMS19 homolog, cytosolic iron-sulfur assembly component; MMS19 nucleotide excision repair homolog; MMS19 nucleotide excision repair homolog (S. cerevisiae); MMS19 nucleotide excision repair protein homolog; MMS19 nucleotide excision repair-like; Mms19l; MMS19-like (MET18 homolog, S. cerevisiae); MMS19-like protein
Gene Symbols MMS19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MMS19. Recombinant protein control fragment (Product #RP-106363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64210
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.