Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMS21 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566946
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6946 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6946 Supplier Bethyl Laboratories, Inc Supplier No. A304129AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

MMS21, also known as NSE2, is a SUMO ligase that in combination with the SMC5/6 complex is required for the prevention of DNA damage induced apoptosis by facilitating DNA repair in human cells. MMS21-dependent sumoylation is integral and important to the cohesion mechanism and mitotic progression; this function appears to be independent of SMC6. MMS21 mediates SUMO attachment to various proteins such as SMC6L1 and TRAX, and possibly the cohesin components RAD21 and STAG2.

Specifications

Antigen MMS21
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NSMCE2
Gene Accession No. Q96MF7
Gene Alias C8orf36, MMS21, NSE2; E3 SUMO-protein ligase NSE2; MMS21 homolog; hMMS21; methyl methanesulfonate sensitivity gene 21; non-SMC element 2 homolog; non-structural maintece of chromosomes element 2 homolog
Gene Symbols NSMCE2
Host Species Rabbit
Immunogen Between 197 and 247
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286053
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.