Learn More
Invitrogen™ MMS22L Polyclonal Antibody

Description
Immunogen sequence: MSQVVPFSQLA DAAADFTLLA MDMPSTAPSD FQPQPVISII QLFGWDDIIC PQVVARYLSH VLQNSTLCEA LSHSGYVSFQ ALTVRSWIRC Highest antigen sequence identity to the following orthologs - mouse 81%, rat 82%.
Specifications
Specifications
| Antigen | MMS22L |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MMS22L |
| Gene Accession No. | Q6ZRQ5 |
| Gene Alias | C6orf167; dJ39B17.2; Methyl methanesulfonate-sensitivity protein 22-like; MMS22 like, DNA repair protein; MMS22L; MMS22-like, DNA repair protein; Protein MMS22-like |
| Gene Symbols | MMS22L |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.