Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMSET / WHSC1 Monoclonal Antibody (29D1), Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 39879

Description
This gene encodes a protein that contains four domains present in other developmental proteins: a PWWP domain, an HMG box, a SET domain, and a PHD-type zinc finger. It is expressed ubiquitously in early development. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene maps to the 165 kb WHS critical region and has also been involved in the chromosomal translocation t(4;14)(p16.3;q32.3) in multiple myelomas. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. Some transcript variants are nonsense-mediated mRNA (NMD) decay candidates, hence not represented as reference sequences.Specifications
MMSET / WHSC1 | |
Monoclonal | |
1 mg/mL | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
O96028 | |
NSD2 | |
This MMSET / WHSC1 antibody was raised against a recombinant protein corresponding to amino acids 1-647 of human MMSET-I. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence | |
29D1 | |
Unconjugated | |
NSD2 | |
5830445G22Rik; 9430010A17Rik; AW555663; C130020C13Rik; D030027O06Rik; D930023B08Rik; Histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; KIAA1090; mKIAA1090; MMSET; multiple myeloma SET domain containing protein type III; Multiple myeloma SET domain-containing protein; NSD2; nuclear receptor binding SET domain protein 2; Nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; REIIBP; RGD1565590; trithorax/ash1-related protein 5; TRX5; WHS; WHSC1; Whsc1l; Wolf-Hirschhorn syndrome candidate 1; Wolf-Hirschhorn syndrome candidate 1 (human); Wolf-Hirschhorn syndrome candidate 1 protein; Wolf-Hirschhorn syndrome candidate 1 protein homolog | |
Mouse | |
Protein G | |
RUO | |
7468 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction