Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMTAG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA561171
Description
Immunogen sequence: MFGSSRGGVR GGQDQFNWED VKTDKQRENY LGNSLMAPVG RWQKGRDLTW YAKGRAPCAG PSREEELAAV REAEREALLA ALGYKNVKK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
MMTAG2 gene ontology (GO) annotations include neutrophil degranulation.
Specifications
MMTAG2 | |
Polyclonal | |
Unconjugated | |
C1orf35 | |
2310033P09Rik; C1orf35; chromosome 1 open reading frame 35; hMMTAG2; MMTAG2; multiple myeloma transforming 2; multiple myeloma tumor-associated protein 2; Multiple myeloma tumor-associated protein 2 homolog; RGD1304587; RIKEN cDNA 2310033P09 gene; similar to RIKEN cDNA 2310033P09 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BU76 | |
C1orf35 | |
Recombinant protein corresponding to Human MMTAG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction