Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMTV Antibody, Novus Biologicals™
SDP

Catalog No. NBP24417905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24417905 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24417905 Supplier Novus Biologicals Supplier No. NBP2441790.05ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMTV Polyclonal specifically detects MMTV in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MMTV
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 66 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, ELISA 1:10000-1:20000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias Envelope glycoprotein gp70, Glycoprotein 36, Glycoprotein 52, Mouse mammary tumor virus (strain GR), Surface protein, Transmembrane protein
Gene Symbols env
Host Species Rabbit
Immunogen MMTV was prepared from whole rabbit serum produced by repeated immunizations with a full length sequence for mouse mammary tumor virus surface protein tagged with His.
Quantity 0.05 mL
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.