Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMTV Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990151
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0151 100 μL
50-199-0152 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0151 Supplier Rockland Immunochemicals Supplier No. 100401P12
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with MMTV from mouse based on a 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

Mouse mammary tumor virus capsid is a polyprotein that is cleaved by aspartyl protease during or after the release of the virion from the plasma membrane. After entering the cell, reverse transcriptase converts the viral dimeric RNA genome into dsDNA in the cytoplasm. Displaying DNA polymerase activity and RNase H activity, this enzyme copies either DNA or RNA templates and cleaves the RNA strand of the RNA-DNA heteroduplex in a partially processive 3' to 5' endonucleasic mode. tRNS binds to the primer at the 5' end of the viral RNA. Reverse transcriptase uses the 3' end of the tRNA primer to perform a short round of RNA-dependent minus-strand DNA synthesis proceeding through the U5 region and ending after the repeated region. This RNA-DNA heteroduplex is digested by the RNase H and hybridizes with the identical R region at the 3' end of the viral RNA. RNase H then digests the RNA template except for a polypurine tract situated at the 5' end of the genome. RNase H probably can proceed both in a polymerase-dependent and a polymerase-independent mode. Reverse transcriptase also performs DNA-directed plus-strand DNA synthesis using the polypurine tract that has not been removed by RNase H as primers. Polypurine tract and tRNA primers are then removed by RNase H and the 3' and 5' ssDNA primer binding site regions hybridize to form a circular dsDNA intermediate. Strand displacement synthesis by reverse transcriptase to the primer binding site and polypurine tract ends produces a blunt ended, linear dsDNA copy of the viral genome that includes long terminal repeats at both ends. Anti-MMTV Antibody is ideal for researchers interested in Cancer, Enzyme, and Viral research.

Specifications

Antigen MMTV
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 77 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen MMTV was prepared from whole rabbit serum produced by repeated immunizations with a full length sequence for mouse mammary tumor virus capsid protein tagged with His.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.