Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MN1 Antibody, Novus Biologicals™
SDP

Catalog No. p-200060682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB408922 100 μg
NB408921 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB408922 Supplier Novus Biologicals Supplier No. NBP306351100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MN1 Polyclonal antibody specifically detects MN1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MN1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500
Formulation PBS, pH7.0
Gene Alias dJ353E16.2, meningioma (disrupted in balanced translocation) 1, meningioma (translocation balanced), meningioma chromosome region, meningioma chromosome region 1, MGCR, MGCR1, MGCR1-PEN, probable tumor suppressor protein MN1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MN1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4330
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.