Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MNAR Recombinant Rabbit Monoclonal Antibody (6H4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538428
Description
This antibody has been tested in direct-ELISA.
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications
MNAR | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 | |
Q8IZL8 | |
PELP1 | |
A synthesized peptide derived from human PELP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
6H4 | |
Unconjugated | |
PELP1 | |
4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27043 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction