Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MNAR Recombinant Rabbit Monoclonal Antibody (9V9A5)
GREENER_CHOICE

Catalog No. PIMA535748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535748 Supplier Invitrogen™ Supplier No. MA535748
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EPEPGLLLEV EEPGTEEERG ADTAPTLAPE ALPSQGEVER EGESPAAGPP PQELVEEEPS APPTLLEEET EDGSDKVQPP PETPAEEEME TETEAEALQE K.

This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNAR
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9V9A5
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PELP1
Gene Accession No. Q56B11, Q8IZL8, Q9DBD5
Gene Alias 4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3
Gene Symbols PELP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human PELP1 (Q8IZL8).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27043, 360552, 75273
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.