Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113425
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113425 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113425 Supplier Invitrogen™ Supplier No. PA5113425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MND1 Polyclonal Antibody for Western Blot, ELISA

Required for proper homologous chromosome pairing and efficient cross-over and intragenic recombination during meiosis. Stimulates both DMC1- and RAD51-mediated homologous strand assimilation, which is required for the resolution of meiotic double-strand breaks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MND1
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4
Gene MND1
Gene Accession No. Q9BWT6
Gene Alias 2610034E18Rik; AI591601; GAJ; homolog of yeast MND1; meiotic nuclear division protein 1 homolog; meiotic nuclear divisions 1; meiotic nuclear divisions 1 homolog; meiotic nuclear divisions 1 homolog (S. cerevisiae); MND1; RGD1309007
Gene Symbols MND1
Host Species Rabbit
Immunogen Recombinant Human Meiotic nuclear division protein 1 homolog protein (2-205AA).
Purification Method Antigen affinity chromatography, Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84057
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.