Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MND1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988024
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8024 Supplier Bioss Supplier No. BS7799R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for proper homologous chromosome pairing and efficient cross-over and intragenic recombination during meiosis. Stimulates both DMC1- and RAD51-mediated homologous strand assimilation, which is required for the resolution of meiotic double-strand breaks.

Specifications

Antigen MND1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MND1
Gene Accession No. Q9BWT6
Gene Alias 2610034E18Rik; AI591601; GAJ; homolog of yeast MND1; Meiotic nuclear division protein 1 homolog; meiotic nuclear divisions 1; meiotic nuclear divisions 1 homolog; meiotic nuclear divisions 1 homolog (S. cerevisiae); Mnd1
Gene Symbols MND1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GAJ/MND1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84057
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.