Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MNDA Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318135AF405
This item is not returnable.
View return policy
Description
MNDA Polyclonal antibody specifically detects MNDA in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
MNDA | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry | |
myeloid cell nuclear differentiation antigen, PYHIN3 | |
Antibody was raised against a peptide corresponding to 15 amino acids near the center of human MNDA. The immunogen is located within 110-160 amino acids of MNDA. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
4332 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction