Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MNDA Antibody (OTI4H6), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272741JF549
Description
MNDA Monoclonal specifically detects MNDA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MNDA | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MNDA | |
Recombinant protein fragment corresponding to amino acids 37-321 of human MNDA (NP_002423) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI4H6 | |
Western Blot, Immunohistochemistry | |
myeloid cell nuclear differentiation antigen, PYHIN3 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
4332.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction