Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MNDA Antibody (OTI4H6), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272741JF646
Description
MNDA Monoclonal specifically detects MNDA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| MNDA | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MNDA | |
| Recombinant protein fragment corresponding to amino acids 37-321 of human MNDA (NP_002423) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI4H6 | |
| Western Blot, Immunohistochemistry | |
| myeloid cell nuclear differentiation antigen, PYHIN3 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 4332.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction