Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MNDA Mouse anti-Human, Alexa Fluor™ 647, Clone: 253 (also known as 253A), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566582
Description
MNDA Monoclonal antibody specifically detects MNDA in Human samples. It is validated for Flow Cytometry,Immunofluorescence.
Specifications
MNDA | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
253 (also known as 253A) | |
Alexa Fluor 647 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction