Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNDA Mouse anti-Human, Clone: OTI4H6, lyophilized, TrueMAB™
SDP

Catalog No. 501668145
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8145 Supplier OriGene Supplier No. CF807178
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation.

Specifications

Antigen MNDA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H6
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MNDA
Gene Accession No. P41218
Gene Alias PYHIN3
Gene Symbols MNDA
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 37-321 of human MNDA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4332
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.