Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MNF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558659 Supplier Invitrogen™ Supplier No. PA558659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NYYKHKYPRP RDTSFSGLSL EEYKLILSTD TLEELKEIDK GMWKKLQEKF APKGPEADDK A Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.

This gene encodes a nucleoid protein localized to the mitochondria inner membrane. The encoded protein affects regulation of insulin secretion, mitochondrial ATP production, and myogenesis through modulation of mitochondrial respiratory chain activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UQCC2
Gene Accession No. Q9BRT2
Gene Alias 2900010M23Rik; bA6B20.2; breast cancer-associated protein SGA-81M; C6orf125; Cbp6; cytochrome B protein synthesis 6 homolog; M19; mitochondrial nucleoid factor 1; Mitochondrial protein M19; MNF1; RGD1306917; ubiquinol-cytochrome c reductase complex assembly factor 2; ubiquinol-cytochrome-c reductase complex assembly factor 2; UQCC2
Gene Symbols UQCC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MNF1. Recombinant protein control fragment (Product #RP-94873).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84300
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.