Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MNI 137
GREENER_CHOICE

Catalog No. 438810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-881-0 10 mg
43-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-881-0 Supplier Tocris Bioscience™ Supplier No. 4388/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective negative allosteric modulator of group II mGlu receptors

Selective negative allosteric modulator of group II mGlu receptors (IC50 values are 8.3 and 12.6 nM for human and rat mGlu2 inhibition of glutamate-induced calcium mobilization). Displays no activity at mGlu1, mGlu4, mGlu5 or mGlu8 receptors in a calcium mobilization assay.

Chemical Identifiers

CAS 946619-21-2
Molecular Formula C15H9BrN4O

Specifications

Quantity 10 mg
Formula Weight 341.16
Purity >98%
Molecular Weight (g/mol) 341.168
Chemical Name or Material MNI 137
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.