Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726051 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6051 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6051 Supplier Proteintech Group Inc Supplier No. 101361AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MKNK1 encodes a predicted 424-amino acid protein containing a sequence typical of the catalytic domain of serine/threonine kinases and a putative N-terminal nuclear localization signal sequence. C-terminal region of MKNK1 was phosphorylated and activated in vivo and in vitro by ERK1 and p38 MAP kinases, but not by JNK/SAPK. The activated ERK is essential for the activation of MKNK1 in insect cells. MKNK1 was activated following stimulation of HeLa cells with a variety of extracellular stimuli. MNK1 bind tightly to the growth factor-regulated MAP kinases, Erk1 and Erk2, and also binds strongly to the stress-activated kinase, p38.

Specifications

Antigen MNK1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MKNK1
Gene Accession No. O08605, Q4G050, Q9BUB5
Gene Alias MKNK1, MNK1
Gene Symbols Mknk1
Host Species Rabbit
Immunogen MNK1 Fusion Protein Ag0184
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17346, 500526, 8569
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.