Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MNK2 Antibody - a Isoform, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260785
Description
MNK2 Polyclonal specifically detects MNK2 in Human, Primate samples. It is validated for Western Blot.Specifications
MNK2 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
EC 2.7.11, EC 2.7.11.1, G protein-coupled receptor kinase 7, GPRK7Putative map kinase interacting kinase, MAP kinase interacting serine/threonine kinase 2, MAP kinase signal-integrating kinase 2, Mnk2, MNK2MAP kinase-interacting serine/threonine-protein kinase 2 | |
Rabbit | |
47 kDa | |
100 μL | |
Primary | |
Human, Primate | |
IgG |
Western Blot | |
Unconjugated | |
10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
MKNK2 | |
Synthetic peptide from the C-terminal region, specific to the human Mnk2a isoform. Accession # Q9HBH9 | |
Antigen Affinity-purified | |
Protein Kinase | |
2872 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction