Learn More
MNS1 Mouse anti-Human, Clone: OTI3E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810322

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MNS1 may play a role in the control of meiotic division and germ cell differentiation through regulation of pairing and recombination during meiosis.Specifications
MNS1 | |
Monoclonal | |
Unconjugated | |
MNS1 | |
SPATA40 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55329 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8NEH6 | |
MNS1 | |
Full length human recombit protein of human MNS1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.