Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MNS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114101 Supplier Invitrogen™ Supplier No. PA5114101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVQENEEKRL QLQNALTQKL EEMLRQREDL EQVRQELYQE EQAEIYKSKL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 93%.

MNS1 may play a role in the control of meiotic division and germ cell differentiation through regulation of pairing and recombination during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MNS1
Gene Accession No. Q8NEH6
Gene Alias AW546487; meiosis specific nuclear structural 1; meiosis-specific nuclear structural 1; meiosis-specific nuclear structural protein 1; Mns1; SPATA40; spermatogenesis associated 40
Gene Symbols MNS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MNS1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55329
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.