Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MNT Recombinant Rabbit Monoclonal Antibody (HL2309)
GREENER_CHOICE

Catalog No. FEMA550486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550486 Supplier Invitrogen™ Supplier No. MA550486
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MNT Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MNT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2309
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MNT
Gene Accession No. Q99583
Gene Alias BHLHD3; Class D basic helix-loop-helix protein 3; MAD6; max binding protein; MAX network transcriptional repressor; max-binding protein MNT; Max-interacting protein; MNT; MNT, MAX dimerization protein; MXD6; Myc antagonist MNT; Protein ROX; Rox
Gene Symbols MNT
Host Species Rabbit
Immunogen Recombinant fragmemt of human MNT.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4335
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.