Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MNX1/HLXB9 Antibody (SR1689), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MNX1/HLXB9 |
---|---|
Clone | SR1689 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
MNX1/HLXB9 Monoclonal antibody specifically detects MNX1/HLXB9 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
MNX1/HLXB9 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
HB9, HLXB9motor neuron and pancreas homeobox protein 1, homeobox HB9, HOXHB9Homeobox protein HB9, motor neuron and pancreas homeobox 1, SCRA1homeo box HB9 | |
A synthesized peptide derived from human MNX1/HLXB9 (Uniprot #: P50219) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1689 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Biology, Neuroscience, Stem Cells | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3110 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title