Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514268
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
The protein encoded by this gene is a component of the Hippo signaling pathway, which controls organ size and tumor growth by enhancing apoptosis. Loss of the encoded protein results in cell proliferation and cancer formation. The encoded protein is also involved in the control of microtubule stability during cytokinesis. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
MOB1 | |
Polyclonal | |
Unconjugated | |
Mob1b | |
1110003E08Rik; 4022402H07Rik; AU015450; B230364F10; C2orf6; MATS1; MATS2; MOB kinase activator 1A; MOB kinase activator 1B; MOB1; Mob1 alpha; mob1 homolog 1A; Mob1 homolog 1B; MOB1 Mps One Binder homolog A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; MOB1, Mps One Binder kinase activator-like 1B; mob1A; Mob1b; MOB4A; MOB4B; Mobk1b; MOBKL1A; MOBKL1B; MOBKL1B/MOB4A; mps one binder kinase activator-like 1A; Mps one binder kinase activator-like 1B; Protein Mob4B | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
55233, 92597 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q7L9L4, Q9H8S9 | |
MOB1A, Mob1b | |
KLH conjugated synthetic peptide between 188-216 amino acids from the C-terminal region of human MOBKL1B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction