Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115585 Supplier Invitrogen™ Supplier No. PA5115585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MOB1A.

The protein encoded by this gene is a component of the Hippo signaling pathway, which controls organ size and tumor growth by enhancing apoptosis. Loss of the encoded protein results in cell proliferation and cancer formation. The encoded protein is also involved in the control of microtubule stability during cytokinesis. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOB1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Mob1b
Gene Accession No. Q7L9L4, Q8BPB0, Q921Y0, Q9H8S9
Gene Alias 1110003E08Rik; 4022402H07Rik; AU015450; B230364F10; C2orf6; MATS1; MATS2; MOB kinase activator 1A; MOB kinase activator 1B; MOB1; Mob1 alpha; mob1 homolog 1A; Mob1 homolog 1B; MOB1 Mps One Binder homolog A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; MOB1, Mps One Binder kinase activator-like 1B; mob1A; Mob1b; MOB4A; MOB4B; Mobk1b; MOBKL1A; MOBKL1B; MOBKL1B/MOB4A; mps one binder kinase activator-like 1A; Mps one binder kinase activator-like 1B; Protein Mob4B
Gene Symbols MOB1A, Mob1b
Host Species Rabbit
Immunogen A synthesized peptide derived from human MOB1B(Accession Q7L9L4), corresponding to amino acid residues M1-E51.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232157, 55233, 68473, 92597
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.