Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOB1A Monoclonal Antibody (2D3D11)
GREENER_CHOICE

Catalog No. PIMA531800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531800 Supplier Invitrogen™ Supplier No. MA531800
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31800 has been tested in indirect ELISA.

The protein encoded by this gene is a component of the Hippo signaling pathway, which controls organ size and tumor growth by enhancing apoptosis. Loss of the encoded protein results in cell proliferation and cancer formation. The encoded protein is also involved in the control of microtubule stability during cytokinesis. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOB1A
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2D3D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene MOB1A
Gene Accession No. Q9H8S9
Gene Alias C2orf6; MATS1; MATS2; MOB kinase activator 1A; MOB kinase activator 1B; MOB1; Mob1 alpha; mob1 homolog 1A; Mob1 homolog 1B; MOB1 Mps One Binder homolog A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; MOB1, Mps One Binder kinase activator-like 1B; mob1A; MOB1B; MOB4A; Mob4B; MOBK1B; MOBKL1A; MOBKL1B; Mps one binder kinase activator-like 1A; mps one binder kinase activator-like 1B
Gene Symbols MOB1A
Host Species Mouse
Immunogen Purified recombinant fragment of human MOB1A (AA: 1-216) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.