Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOB3A/MOB3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575725
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MOBKL2A (Mps one binder kinase activator-like 2A), also known as MOB-LAK or MOB3A, is a 217 amino acid protein that regulates kinase activity. A member of the MOB1/phocein family, MOBKL2A is encoded by a gene that maps to human chromosome 19. Consisting of around 63 million bases with over 1,400 genes, chromosome 19 makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin superfamily members including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family, and Fcα receptors. Key genes for eye color and hair color also map to chromosome 19. Peutz-Jeghers syndrome, spinocerebellar ataxia type 6, the stroke disorder CADASIL, hypercholesterolemia and insulin-dependent diabetes have been linked to chromosome 19.
Specifications
MOB3A/MOB3B | |
Polyclonal | |
Unconjugated | |
MOB3B | |
5330417K06Rik; 8430436F23Rik; A430018A01Rik; A630029F06; AV218468; C9orf35; MOB kinase activator 3A; MOB kinase activator 3B; MOB kinase activator-like 2B; MOB LAK; MOB1 (Mps One Binder) kinase activator-like 2B; mob1 homolog 2A; Mob1 homolog 2b; MOB1, Mps One Binder kinase activator-like 2A; MOB1, Mps One Binder kinase activator-like 2B; MOB1C; MOB1D; Mob3a; MOB3B; MOBKL2A; Mobkl2b; moblak; MOB-LAK; monopolar spindle 1 binding, MOB1, domain containing; mps one binder kinase activator-like 2A; Mps one binder kinase activator-like 2B; RGD1560696 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126308, 208228, 214944, 362833, 366352, 79817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TA1, Q8BSU7, Q8VE04, Q96BX8 | |
MOB3A, MOB3B | |
Synthetic peptide corresponding to residues in Human MOB3A/MOB3B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction