Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560622 Supplier Invitrogen™ Supplier No. PA560622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQFCLELNGL AVKLQSECHP DTCTQMTATE QWIFLCAAHK TPKECPAIDY TRHTLDGAAC LLNSNKYFPS RVSIKES Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene was identified based on its similarity with the mouse counterpart. Studies of the mouse counterpart suggest that the expression of this gene may be regulated during oocyte maturation and preimplantation following zygotic gene activation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. Naturally occurring read-through transcription occurs between this locus and the neighboring locus HSPE1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOB4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOB4
Gene Accession No. Q9Y3A3
Gene Alias 2610109B12Rik; 2C4D; CGI-95; Class II mMOB1; MOB family member 4, phocein; MOB1; Mob1 homolog 3; MOB1, Mps One Binder kinase activator-like 3; mob3; MOB4; Mobkl1; Mobkl3; MOB-like protein phocein; mps one binder kinase activator-like 3; Phocein; phocein, Mob-like protein; PHOCN; PREI3; Preimplantation protein 3
Gene Symbols MOB4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MOB4. Recombinant protein control fragment (Product #RP-99626).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25843
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.