Learn More
Invitrogen™ MOB4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592261
Description
Immunogen sequence: MVMAEGTAVL RRNRPGTKAQ DFYNWPDESF DEMDSTLAVQ QYIQQNIRAD CSNIDKILEP PEGQDEGVWK YEHLRQFCLE LNGLAVKLQS ECHPDTCTQM TATEQWIFLC AAHKTPKECP AIDYTRHTLD GAACLLNSNK YFPSRVSIKE SSVAKLGSVC RRIYRIFSHA YFHHRQIFDE YENETFLCHR FTKFVMKYNL MSKDNLIVPI LEEEVQNSVS GESEA; Positive Samples: A-549, MCF-7, Mouse brain, Mouse heart, Mouse kidney, Rat spleen, Rat brain; Cellular Location: Cytoplasm, Golgi apparatus, Golgi stack membrane, Membrane, Peripheral membrane protein, perinuclear region.
This gene was identified based on its similarity with the mouse counterpart. Studies of the mouse counterpart suggest that the expression of this gene may be regulated during oocyte maturation and preimplantation following zygotic gene activation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. Naturally occurring read-through transcription occurs between this locus and the neighboring locus HSPE1.
Specifications
MOB4 | |
Polyclonal | |
Unconjugated | |
MOB4 | |
2610109B12Rik; 2C4D; CGI-95; Class II mMOB1; MOB family member 4, phocein; MOB1; Mob1 homolog 3; MOB1, Mps One Binder kinase activator-like 3; mob3; MOB4; Mobkl1; Mobkl3; MOB-like protein phocein; mps one binder kinase activator-like 3; Phocein; phocein, Mob-like protein; PHOCN; PREI3; Preimplantation protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
171050, 19070, 25843 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6PEB6, Q9QYW3, Q9Y3A3 | |
MOB4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-225 of human MOB4 (NP_0562022). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.