Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOB4A Antibody (OTI3G9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272744F
Description
MOB4A Monoclonal specifically detects MOB4A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
MOB4A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MOB1B | |
Full length recombinant protein of human MOB4A (NP_775739) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunohistochemistry | |
OTI3G9 | |
Immunohistochemistry | |
MATS2, MGC33910, Mob1 homolog 1A, MOB1, Mps One Binder kinase activator-like 1A (yeast), Mob1A, Mob1Bmps one binder kinase activator-like 1A, MOB4Amob1A, Protein Mob4A | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
92597.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction