Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOB4A/MOBKL1B Antibody (01), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306512IR
Description
MOB4A/MOBKL1B Monoclonal antibody specifically detects MOB4A/MOBKL1B in Human samples. It is validated for ImmunofluorescenceSpecifications
MOB4A/MOBKL1B | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
92597 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
01 | |
Immunocytochemistry/Immunofluorescence | |
ATS2;MATS2;MOB kinase activator 1B;MOB1B;MOB4A;MOBKL1A | |
This antibody was obtained from a mouse immunized with purified, recombinant Human MOB4A/MOBKL1B (Q7L9L4; Met1-Arg216). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction