Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOB4A Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA529426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529426 Supplier Invitrogen™ Supplier No. MA529426
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MOB1B/MOBKL1A.

MOB4A is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Mob4A regulates NDR1 and NDR2 kinase activity, widely expressed nuclear serine-threonine kinases that have been implicated in cell proliferation and/or tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOB4A
Applications Immunocytochemistry
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MOB1B
Gene Accession No. Q7L9L4
Gene Alias 1110003E08Rik; AU015450; B230364F10; MATS2; MGC33910; MOB kinase activator 1B; mob1 homolog 1A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; mob1A; MOB1B; MOB4A; Mobkl1a; mps one binder kinase activator-like 1A
Gene Symbols MOB1B
Host Species Mouse
Immunogen Recombinant Human MOB1B/MOBKL1A Protein (Met1-Arg216).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92597
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.