Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL1A Monoclonal Antibody (OTI3G9), TrueMAB™, OriGene
SDP

Catalog No. 502542348
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2348 30 μL
50-167-2506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2348 Supplier OriGene Supplier No. TA501363S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOBKL1A Monoclonal antibody specifically detects MOBKL1A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MOBKL1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MOB1B
Gene Accession No. Q7L9L4
Gene Alias 1110003E08Rik; AU015450; B230364F10; MATS2; MGC33910; MOB kinase activator 1B; mob1 homolog 1A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; mob1A; MOB1B; MOB4A; Mobkl1a; mps one binder kinase activator-like 1A
Host Species Mouse
Immunogen Full length human recombinant protein of human MOBKL1A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92597
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.