Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOBKL2B Antibody (OTI6C6), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272748MFV450
Description
MOBKL2B Monoclonal antibody specifically detects MOBKL2B in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
MOBKL2B | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
79817 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI6C6 | |
Flow Cytometry | |
Em:AL163192.1, FLJ13204, FLJ23916, Mob1 homolog 2b, MOB1, Mps One Binder kinase activator-like 2B (yeast), MOB3BMGC32960, monopolar spindle 1 binding, MOB1, domain containing, mps one binder kinase activator-like 2B, Protein Mob3B | |
Full length human recombinant protein of human MOBKL2B (NP_079037) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction