Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL2B Monoclonal Antibody (OTI6C6), TrueMAB™, OriGene
SDP

Catalog No. 501672642
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2642 100 μL
50-254-2235 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2642 Supplier OriGene Supplier No. TA501501
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene shares similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. This gene is located on the opposite strand as the interferon kappa precursor (IFNK) gene.

Specifications

Antigen MOBKL2B
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI6C6
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MOB3B
Gene Accession No. Q86TA1
Gene Alias C9orf35, MOB1D, MOBKL2B
Gene Symbols MOB3B
Host Species Mouse
Immunogen Full length human recombit protein of human MOBKL2B produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79817
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.