Learn More
MOBKL2B Mouse anti-Human, Clone: OTI1G1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501499

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene shares similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. This gene is located on the opposite strand as the interferon kappa precursor (IFNK) gene.Specifications
MOBKL2B | |
Monoclonal | |
Unconjugated | |
MOB3B | |
C9orf35, MOB1D, MOBKL2B | |
Mouse | |
Affinity Chromatography | |
RUO | |
79817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86TA1 | |
MOB3B | |
Full length human recombit protein of human MOBKL2B produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.