Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOBKL3 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299981AF594
Description
MOBKL3 Polyclonal specifically detects MOBKL3 in Human samples. It is validated for Western Blot, ELISA.Specifications
MOBKL3 | |
Polyclonal | |
Western Blot, ELISA | |
2C4D, CGI-95, Class II mMOB1, MOB1, MOB1, Mps One Binder kinase activator-like 3 (yeast), MOB1DKFZP564M112, Mob3, MOB3MGC12264, MOB4 MOB family member 4, phocein, MOBKL3, phocein, PHOCN, PREI3, PREI3mps one binder kinase activator-like 3, Preimplantation protein 3Mob1 homolog 3,2C4DCGI-95 | |
Produced in rabbits immunized with purified, recombinant MOBKL3 (Q9Y3A3-1; Val 2-Ala 225). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
25843 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction