Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100618 Supplier Thermo Scientific Supplier No. PA5100618
Add to Cart
Add to Cart

MOBP Polyclonal Antibody

MOBP Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MOBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mobp
Gene Accession No. Q13875, Q63327, Q9D2P8
Gene Alias AI835053; AI839848; Mobp; MOBP155; MOBP170; MOBP69; MOBP73; Mobp81; Mobp81p; myelin associated oligodendrocyte basic protein; myelin-associated oligodendrocyte basic protein; myelin-associated oligodendrocytic basic protein; Myelin-associated/Oligodendrocytic Basic Protein-81; R74645; RNMOBP81P; Unknown (protein for MGC:128384)
Gene Symbols Mobp
Host Species Rabbit
Immunogen A synthesized peptide derived from human MOBP(Accession Q13875), corresponding to amino acid residues R118-P168.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17433, 25037, 4336
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.