Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557179
Description
Immunogen sequence: SQKPAKEGPR LSKNQKYSEH FSIHCCPPFT FLNSKKEIVD RKYSICKSGC FYQKKEEDWI CCACQKTR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
MOBP may play a role in compacting or stabilizing the myelin sheath, possibly by binding the negatively charged acidic phospholipids of the cytoplasmic membrane.
Specifications
MOBP | |
Polyclonal | |
Unconjugated | |
Mobp | |
AI835053; AI839848; Mobp; MOBP155; MOBP170; MOBP69; MOBP73; Mobp81; Mobp81p; myelin associated oligodendrocyte basic protein; myelin-associated oligodendrocyte basic protein; myelin-associated oligodendrocytic basic protein; Myelin-associated/Oligodendrocytic Basic Protein-81; R74645; RNMOBP81P; Unknown (protein for MGC:128384) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13875 | |
Mobp | |
Recombinant protein corresponding to Human MOBP. Recombinant protein control fragment (Product #RP-96435). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction