Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOCOS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530662
Description
Recommended positive controls: 293T, HepG2. Predicted reactivity: Mouse (83%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MOCOS sulfurates the molybdenum cofactor of xanthine dehydrogenase (XDH), which is required for their enzymatic activities (Ichida et al., 2001).
Specifications
MOCOS | |
Polyclonal | |
Unconjugated | |
MOCOS | |
1110018O12Rik; HMCS; MCS; moCo sulfurase; MOCOS; Molybdenum cofactor sulfurase; molybdenum cofactor sulfurtransferase; MOS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55034 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96EN8 | |
MOCOS | |
Recombinant protein encompassing a sequence within the N-terminus region of human MOCOS. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction